Learn More
Medchemexpress LLC N-(2-hydroxyethyl)maleimide | 1585-90-6 | MFCD00465266 | 99.2% | 141.12 g/mol | C6H7NO3 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0175885G
Encompass_Preferred
N-(2-hydroxyethyl)maleimide is a maleimide-based cross-linker bearing a 2-hydroxyethyl substituent. It provides a thiol-reactive maleimide and a hydroxyl handle for downstream modification, commonly used in chemical synthesis and bioconjugation research.
  • Reactive maleimide moiety for thiol-selective conjugation.
  • Hydroxyl functional group enables further derivatization.
  • Low molecular weight (141.12 g/mol) for minimal steric hindrance.
  • High purity suitable for research applications (≈99.2%).
  • Available in small lab-scale pack sizes for synthetic workflows.
Catalog No. 50-003-00426