Learn More
Medchemexpress LLC N-[2-methoxy-4-(4-methylpiperazin-1-yl)phenyl]-4-(1-methylpyrrolopyridin-3-yl)pyrimidin-2-amine | 1594092-37-1 | >98.0% | 1 ML

Supplier: Medchemexpress LLC HY1227710MM 1ML
AZ191 is a potent, selective small-molecule inhibitor of DYRK1B (IC50 = 17 nM) supplied for research use. It is typically provided as a concentrated solution (10 mM, 1 mL); refer to the datasheet for detailed solubility and storage guidance.
- Potent DYRK1B inhibitor with IC50 of 17 nM.
- High purity: ≥98% (HPLC).
- Chemical identifiers: CAS 1594092-37-1; formula C24H27N7O; MW 429.52 g/mol.
- Soluble in DMSO (≥30 mg/mL) and can be formulated for in vivo use.
- Storage recommendations: powder -20°C (up to 3 years); in solvent -80°C for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.