Learn More
Cayman Chemical N 2hydroxyethylNaphthalIm 1g
SDP
Supplier:  Cayman Chemical 175671 G
Encompass_Preferred
A fluorescent probe; used as a precursor for protection of amine groups; used to detect nucleic acids and their precursors, which quench its fluorescence; is electroactive; ex/em = 330-333 and 344-347/366-378 nm in solvents of varying polarities
Catalog No. 50-296-6323