Learn More
Cayman Chemical N 2PocethylbetulIn amIde 1mg
SDP
Supplier:  Cayman Chemical 330701 MG
Encompass_Preferred
An alkyne derivative of betulin; can be used for click chemistry applications
Catalog No. 50-294-6618