Learn More
Medchemexpress LLC N-[(2s)-1-[[(2s)-5-(carbamoylamino)-1-[4-(hydroxymethyl)anilino]-1-oxopentan-2-yl]amino]-3-methyl-1-oxobutan-2-yl]-6-(2,5-dioxopyrrol-1-yl)hexanamide | 159857-80-4 | MFCD26142966 | 99.9% | 572.65 | C28H40N6O7 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY787385MG
Encompass_Preferred
MC-Val-Cit-PAB is a protease-cleavable valine-citrulline-PAB linker used as an intermediate in the synthesis of antibody-drug conjugates. It is commonly incorporated into VcMMAE-type drug-linker constructs where the cytotoxic payload is a tubulin inhibitor.
  • Cleavable peptide linker suitable for ADC conjugation.
  • Used as an intermediate for VcMMAE and related drug-linker conjugates.
  • High purity for synthetic and research applications.
  • Solid form, convenient for handling and storage.
  • Stable when stored sealed away from moisture and light; follow recommended temperature conditions.
  • Identifying data: CAS 159857-80-4, molecular formula C28H40N6O7, molecular weight 572.65.
Catalog No. 50-002-03535