Learn More
Medchemexpress LLC N-[3-(1H-benzimidazol-2-yl)-4-chlorophenyl]pyridine-3-carboxamide | 496793-75-0 | 99.9% | 100 MG

Supplier: Medchemexpress LLC HY122024100MG
AZSMO-23 (CAS 496793-75-0) is a small-molecule activator of the hERG (Kv11.1) potassium channel used for in vitro electrophysiology and pharmacology research. It activates wild-type hERG pre-pulse and tail currents with reported EC50 values of 28.6 μM and 11.2 μM, respectively. Supplied as an off-white to light yellow solid with high purity and defined storage recommendations.
- High purity: 99.9%.
- Molecular formula C19H13ClN4O; molecular weight 348.79 g/mol.
- Potent hERG K+ channel activator; EC50 28.6 μM (pre-pulse), 11.2 μM (tail).
- Soluble in DMSO (~5.83 mg/mL) with ultrasonic warming recommended.
- Appearance: off-white to light yellow solid.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.