Learn More
Medchemexpress LLC N-[3-(4-cyanophenyl)phenyl]-N-(pyridin-3-ylmethyl)ethanesulfonamide | 353235-01-5 | 99.7% | 377.46 g/mol | C21H19N3O2S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY107505A5MG
Encompass_Preferred
CBiPES is a small-molecule research compound that acts as a positive allosteric modulator (PAM) of the metabotropic glutamate receptor 2 (mGlu2). It is intended for in vitro and preclinical research applications, and is supplied with analytical data confirming identity and high purity.
Catalog No. 50-002-41269