Learn More
Medchemexpress LLC N-(3,4-difluorophenyl)-1,3,5-trimethyl-4-[2-oxo-2-(prop-2-ynylamino)acetyl]pyrrole-2-carboxamide | 2133017-36-2 | 98.4% | 373.35 g/mol | C19H17F2N3O3 | 25MG
SDP
Supplier:  Medchemexpress LLC HY12461425MG
Encompass_Preferred
GLP-26 is a small-molecule hepatitis B virus (HBV) capsid assembly modulator for research use. It inhibits HBV DNA replication in cellular assays, reduces cccDNA, and lowers secretion of viral antigens. The compound is a solid with good DMSO solubility and defined storage recommendations suitable for laboratory handling.
Catalog No. 50-002-35090