Learn More
Medchemexpress LLC N'-[3,5-bis(trifluoromethyl)phenyl]-N-[4-bromo-2-(2H-tetrazol-5-yl-phenyl]thiourea | 956014-19-0 | MFCD25563257 | 99.5% | C16H9BrF6N6S | 10MG
SDP
Supplier:  Medchemexpress LLC HY1310310MG
Encompass_Preferred
NS 11021 is a small-molecule activator of large-conductance, calcium-activated potassium (BKCa/KCa1.1) channels used in electrophysiology and pharmacology research to increase channel open probability and probe BK-mediated effects on cellular excitability.
  • Potent and selective activator of large-conductance Ca2+-activated K+ channels.
  • High purity (~99.5%) suitable for in vitro studies.
  • Soluble in DMSO at high concentration for easy dosing.
  • Powder stable at -20°C for extended storage.
  • Available in multiple small-scale pack sizes for research workflows.
Catalog No. 50-002-61777