Learn More
Medchemexpress LLC N-[(3-(anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline hydrochloride | 63857-00-1 | MFCD00191759 | 96.5% | 359.29 g/mol | C20H20Cl2N2 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW019306100G
Encompass_Preferred
N-[(3-(Anilinomethylene)-2-chloro-1-cyclohexen-1-yl)methylene]aniline hydrochloride is an organic intermediate used as a building block for the synthesis of cyanine-containing dyes and related compounds. Supplied as a solid (reported dark purple to black), it is intended for research and chemical synthesis workflows that require an anilinomethylene-substituted cyclohexenyl scaffold.
  • Used as a building block for cyanine dye synthesis.
  • Solid form facilitates handling and storage.
  • High purity suitable for research applications.
  • Available in multiple pack sizes for scalable synthesis.
  • Molecular weight 359.29 g/mol; formula C20H20Cl2N2.
Catalog No. 50-003-01370