Learn More
Medchemexpress LLC N-(3-chloro-4-methylphenyl)-3-(3-((1-(2,6-dioxopiperidin-3-yl)-2,5-dioxo-2,5-dihydro-1H- | 2438124-79-7 | 98.6% | 494.93 g/mol | C25H23ClN4O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1457765MG
Encompass_Preferred
ALV1 is a small-molecule molecular glue degrader that induces proteasomal degradation of Ikaros (IKZF1) and Helios (IKZF2) via binding to cereblon (CRBN). It is supplied as a high-purity solid for research use.
  • Potent degrader of Ikaros and Helios (DC50 2.5 nM and 10.3 nM, respectively).
  • Binds cereblon with IC50 of 0.55 μM.
  • High purity solid, light yellow to green-yellow appearance.
  • Soluble in DMSO (50 mg/mL); in vivo formulation examples available.
  • Stable when stored as powder at -20°C (3 years) or 4°C (2 years).
Catalog No. 50-002-21146