Learn More
Medchemexpress LLC N-(3-chlorophenyl)-8-[[1-(4-methyl-2-nitrophenyl)-1H-1,2,3-triazol-4-yl]methoxy]-2-quinazolinamine | 2587177-94-2 | 98.3% | C24H18ClN7O3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13197810MG
Encompass_Preferred
DB18 is a small-molecule inhibitor of CDC2-like kinases (CLKs) with potent, selective activity and reported anti-tumor effects. The compound exhibits low-nanomolar inhibition of CLK isoforms and is supplied as a high-purity solid for research applications.
  • Potent, selective inhibitor of CDC2-like kinases (CLKs).
  • Low-nanomolar IC50 (10-30 nM) for CLK1, CLK2, and CLK4.
  • Demonstrated anti-tumor activity in preclinical studies.
  • High purity suitable for biochemical assays.
  • Soluble in DMSO at 50 mg/mL with ultrasonic warming.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-61055