Learn More
Medchemexpress LLC N-(3-hydroxyphenyl)-adenosine 2',3',5'-triacetate | 1221412-23-2 | 97.7% | 485.45 g/mol | C22H23N5O8 | 5 MG

Supplier: Medchemexpress LLC HY1416455MG
IMM-H007 is an orally active small-molecule activator of AMPK and an antagonist of TGFβ1, supplied for research use in studies of metabolic regulation and fibrotic signaling pathways.
- Orally active AMPK activator and TGFβ1 antagonist.
- High purity (97.69%) white to off-white solid.
- Molecular formula C22H23N5O8; molecular weight 485.45 g/mol.
- Soluble in DMSO at 125 mg/mL; ultrasonic assistance recommended.
- Powder stable at -20°C for up to 3 years; in solution store at -80°C to maximize stability.
- Suitable for biochemical and pharmacological research into AMPK and TGFβ1 pathways.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.