Learn More
Medchemexpress LLC N-(3-indolylacetyl)-L-alanine | 57105-39-2 | MFCD00050395 | 99.9% | 246.27 g/mol | C13H14N2O3 | 100 MG

Supplier: Medchemexpress LLC HYW141858100MG
N-(3-Indolylacetyl)-L-alanine is an indole-acetylated amino acid used as a biochemical research reagent, particularly in studies of auxin-related pathways and amino acid derivatives. Supplied as a high-purity solid, it serves as an analytical standard and a tool compound for plant physiology and biochemical assays.
- Purity 99.9%.
- White to off-white solid physical form.
- Molecular formula C13H14N2O3; molecular weight 246.27 g/mol.
- Suitable for auxin-related and amino acid derivative research.
- Recommended storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.