Learn More
Medchemexpress LLC N-(3-Methoxybenzyl)-(9Z,12Z,15Z)-octadecatrienamide | 883715-23-9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN770210MG
Encompass_Preferred
N-(3-Methoxybenzyl)-(9Z,12Z,15Z)-octadecatrienamide is a macamide isolated from Maca (Lepidium meyenii Walp.). It induces mesenchymal stem cells osteogenic differentiation and consequent bone formation by activating the canonical Wnt/β‐catenin signaling pathway, making it suitable for osteoporosis research.
  • Isolated from Maca (Lepidium meyenii Walp.)
  • Induces mesenchymal stem cells osteogenic differentiation
  • Promotes bone formation via the canonical Wnt/β-catenin signaling pathway
  • Useful for osteoporosis research
Catalog No. 50-004-17303