Learn More
Medchemexpress LLC N-(3-Methoxybenzyl)-(9Z,12Z,15Z)-octadecatrienamide | 883715-23-9 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HYN770210MM/1ML
Encompass_Preferred
N-(3-Methoxybenzyl)-(9Z,12Z,15Z)-octadecatrienamide is a macamide isolated from Maca (Lepidium meyenii Walp.). It can be used for research related to osteoporosis.
  • Induces mesenchymal stem cells osteogenic differentiation.
  • Promotes bone formation.
  • Activates the canonical Wnt/β-catenin signaling pathway.
Catalog No. 50-004-17838