Learn More
Medchemexpress LLC N-(3-Methoxybenzyl)-(9Z,12Z,15Z)-octadecatrienamide | 883715-23-9 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN77025MG
Encompass_Preferred
N-(3-Methoxybenzyl)-(9Z,12Z,15Z)-octadecatrienamide is a macamide isolated from Maca (Lepidium meyenii Walp.). This product induces mesenchymal stem cells osteogenic differentiation and subsequent bone formation by activating the canonical Wnt/β-catenin signaling pathway. It can be utilized for research pertaining to osteoporosis.
  • Isolated from Maca (Lepidium meyenii Walp.).
  • Induces mesenchymal stem cells osteogenic differentiation.
  • Promotes bone formation.
  • Activates the canonical Wnt/β-catenin signaling pathway.
  • Useful for osteoporosis research.
  • Targets Wnt and β-catenin.
  • Involves the Stem Cell/Wnt pathway.
Catalog No. 50-004-17839