Learn More
Medchemexpress LLC N-(3-Methoxybenzyl)Palmitamide | 847361-96-0 | 1 MG

Supplier: Medchemexpress LLC HYN24281MG
N-(3-Methoxybenzyl)Palmitamide is a promising inhibitor of FAAH, used for the treatment of pain, inflammation, and CNS degenerative disorders.
- Purity: 99.96%
- Appearance: Solid
- Color: White to off-white
- Initial source: Plants other families
- For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.