Learn More
Medchemexpress LLC N-(3-Methoxybenzyl)Palmitamide | 847361-96-0 | 5 MG

Supplier: Medchemexpress LLC HYN24285MG
N-(3-Methoxybenzyl)Palmitamide is a promising inhibitor of FAAH (Fatty Acid Amide Hydrolase) intended for the treatment of pain, inflammation, and central nervous system (CNS) degenerative disorders. It is for research use only and not sold to patients.
- Purity of 99.96% ensures reliable experimental results.
- Molecular weight: 375.59
- Formula: C24H41NO2
- Appearance: Solid
- Color: White to off-white
- Structure classification: Others
- Initial source: Plants (other families)
- Soluble in DMSO at 12.5 mg/mL (33.28 mM) for in vitro applications.
- Potential therapeutic applications in research related to pain, inflammation, and CNS degenerative disorders.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.