Learn More
Medchemexpress LLC N-(3-methoxybenzyl)palmitamide | 847361-96-0 | MFCD30722444 | 100.0% | 375.59 g·mol⁻1 | C24H41NO2 | 10 MG

Supplier: Medchemexpress LLC HYN242810MG
N-(3-methoxybenzyl)palmitamide is a research-grade fatty acid amide hydrolase (FAAH) inhibitor used in preclinical studies of pain, inflammation, and neurodegenerative disorders. It is supplied as a purified small-molecule for biochemical and cell-based assays.
- Chemical formula C24H41NO2.
- Molecular weight 375.59 g·mol⁻1.
- CAS number 847361-96-0.
- Target FAAH (fatty acid amide hydrolase).
- Purity 99.96% (supplier specified).
- Available as a 10 mg vial for laboratory use.
- Intended for research use only; not for human or animal therapeutic use.
- Applications include biochemical assays and cellular studies of endocannabinoid signaling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.