Learn More
Medchemexpress LLC N-3-oxo-dodecanoyl-L 50mg | 168982-69-2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY114544A50MG
Encompass_Preferred
N-3-oxo-dodecanoyl-L-homoserine lactone is a bacterial quorum-sensing signaling molecule used in research on microbial communication, host-pathogen interactions, and immunomodulatory pathways. It is supplied as a purified research-grade compound for in vitro biochemical and cell-based assays.
  • Used to study bacterial quorum sensing and signaling pathways
  • High purity suitable for biochemical and cell-based assays
  • Stable solid form for easy handling and storage
  • Available in small-scale research quantities
  • Includes analytical documentation (COA, data sheet, SDS)
Catalog No. 50-003-07867