Learn More
AdipoGen N-(3-Oxododecanoyl)-L-homoserine lactone, 168982-69-2, MFCD08275140, 10 mg
SDP
Supplier:  AdipoGen CDXO0031M010
Encompass_Preferred
MF: C16H27NO4, MW: 297.39, ≥98%, Appearance: White powder. Quorum sensing agent.
Catalog No. 50-595-897