Learn More
Medchemexpress LLC N-(3-Oxooctanoyl)-DL-homoserine lactone | 106983-27-1 | 99.7% | 241.28 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11380110MG
Encompass_Preferred
N-(3-Oxooctanoyl)-DL-homoserine lactone is an N-Acyl homoserine lactone (AHL) derived from gram-negative bacteria. This compound exhibits stereochemistry-dependent growth regulatory activity, particularly affecting root development.
  • Member of N-Acyl homoserine lactone (AHL) family.
  • Derived from gram-negative bacteria.
  • Exhibits stereochemistry-dependent growth regulatory activity.
  • Increases sprouting of roots (S enantiomer shown to be more effective).
  • Affects root cell elongation.
  • Intended for research use only.
Catalog No. 50-004-35834