Learn More
Apexbio Technology LLC N-(3-pyridyl)-Indomethacin amide 261766-29-4 10mg
Supplier: Apexbio Technology LLC C562910
N-3-pyridyl-Indomethacin amide is a small-molecule inhibitor targeting cyclooxygenase-2 (COX-2) It is designed to selectively inhibit the COX-2 isoform thereby modulating inflammatory responses mediated by this enzyme N-3-pyridyl-Indomethacin amide exerts its biological activity primarily through reversible inhibition of COX-2 In studies with recombinant human COX-2 N-3-pyridyl-Indomethacin amide demonstrates potent inhibition with an IC50 value of approximately 0 052 M Based on these pharmacological properties N-3-pyridyl-Indomethacin amide holds research potential in the investigation of COX-2-related pathways in inflammation pain and associated cellular signaling events