Learn More
Cayman Chemical N 3hydroxy7cIs tetradecen 5mg
SDP
Supplier:  Cayman Chemical 90029395 MG
Encompass_Preferred
An AHL involved in quorum sensing; did not affect cytokine or antibody production in mouse and human leukocyte immunoassays using LPS-stimulated macrophages
Catalog No. 50-289-5993