Learn More
Medchemexpress LLC N-[[4-(1,1-dimethylethyl)phenyl]methyl]-N-methyl-L-leucyl-N-(1,1-dimethylethyl)-O-(phenylmethyl)-L-t | 217171-01-2 | MFCD12828777 | 98.9% | 599.85 g·mol⁻1 | C38H53N3O3 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10331850MG
Encompass_Preferred
PD173212 is a selective N-type voltage-sensitive calcium channel (VSCC) blocker used in electrophysiology and neuropharmacology research. It shows an IC50 of 36 nM in IMR-32 assays and is supplied as a purified research chemical.
  • Selective N-type VSCC blocker (IC50 36 nM in IMR-32 assays).
  • High purity (~98.9%) suitable for research applications.
  • Molecular formula C38H53N3O3 and molecular weight 599.85 g·mol⁻1.
  • Supplied as a 50 MG research pack for laboratory use.
  • Commonly used in electrophysiology and neuropharmacology studies.
Catalog No. 50-002-44942