Learn More
Medchemexpress LLC N-(4-(2-phenyl-4-(p-tolyl)piperazine-1-carbonyl)phenyl)acetamide | 1629166-02-4 | 99.6% | 413.51 g/mol | C26H27N3O2 | 100 MG

Supplier: Medchemexpress LLC HY101832100MG
SBC-110736 is a small-molecule inhibitor of proprotein convertase subtilisin/kexin type 9 (PCSK9) supplied as a white to off-white solid for research use. It has formula C26H27N3O2 and a molecular weight of 413.51 g/mol and is used in preclinical studies targeting PCSK9-related pathways.
- High purity (≈99.6%).
- White to off-white solid suitable for handling and formulation.
- Molecular weight 413.51 g/mol and formula C26H27N3O2.
- Soluble in DMSO for stock solution preparation.
- Suitable for preclinical biochemical and in vivo studies of PCSK9 modulation.
- Available in multiple pack sizes, including 100 MG.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.