Learn More
Medchemexpress LLC N-(4-(2-tert-butylphenyl)sulfonylphenyl)-2,3,4-trihydroxy-5-(2-isopropylbenzyl)benzamide | 877877-35-5 | 99.5% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY120205MG
Encompass_Preferred
TW-37 is a small-molecule BH3 mimetic that inhibits Bcl-2 family proteins and induces apoptosis in target cells. It is characterized by biochemical Ki values against Mcl-1, Bcl-2, and Bcl-xL, and is supplied as a high-purity solid for research applications.
  • Potent inhibitor of Bcl-2 family proteins, including Mcl-1, Bcl-2, and Bcl-xL.
  • Reported Ki values: Mcl-1 260 nM, Bcl-2 290 nM, Bcl-xL 1110 nM.
  • High purity (99.5%) suitable for biochemical and cell-based assays.
  • Molecular formula C33H35NO6S and molecular weight 573.7 g/mol.
  • Available as a small solid package or as DMSO solutions for immediate use.
Catalog No. 50-002-33223