Learn More
Medchemexpress LLC N-[4-[3,5-bis(trifluoromethyl)-1H-pyrazol-1-yl]phenyl]-4-methyl-1,2,3-thiadiazole-5-carboxamide | 223499-30-7 | MFCD00220976 | 99.4% | 421.32 g/mol | C15H9F6N5OS | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10083125MG
Encompass_Preferred
YM-58483 (BTP2) is a pyrazole-derived small molecule used in research to inhibit calcium release-activated calcium (CRAC) channels and block store-operated Ca2+ entry (SOCE). It is applied in cellular and immunology studies to reduce sustained calcium influx and downstream Ca2+-dependent signaling and cytokine production.
  • Selective CRAC channel and SOCE inhibitor.
  • Potent activity in cellular assays (IC50 ≈ 100 nM).
  • High-purity solid suitable for research use.
  • Available in small-scale quantities for laboratory experiments.
  • Characterized by CAS number, formula, and molecular weight for traceability.
Catalog No. 50-002-11473