Learn More
Medchemexpress LLC N-[4-(3-amino-hexahydro-dioxoimidazo-pteridin-yl)benzoyl]-L-glutamic acid | 10538-99-5 | MFCD29920315 | 98.0% | 471.42 g·mol⁻¹ | C20H21N7O7 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10194310MM 1ML
Encompass_Preferred
LY 345899 is a folate-analog inhibitor of methylenetetrahydrofolate dehydrogenase enzymes (MTHFD1 and MTHFD2), supplied as a high-purity research compound. It is provided as a solid and as a 10 mM solution in DMSO (1 mL), and is used for biochemical and cellular studies of one-carbon metabolism and oncology research.
  • Inhibits methylenetetrahydrofolate dehydrogenase enzymes (MTHFD1 and MTHFD2).
  • Offered at high purity (≥98.0%).
  • Available as a 10 mM solution in DMSO (1 mL) and as solids in multiple mg sizes.
  • Stable with recommended storage: powder at -20°C, in solvent at -80°C for long-term storage.
  • Suitable for biochemical and cellular research into one-carbon metabolism and oncology.
Catalog No. 50-002-48911