Learn More
Medchemexpress LLC N-[4-(3-bromoanilino)quinazolin-6-yl]prop-2-enamide | 194423-15-9 | 98.6% | C17H13BrN4O | 10MG
SDP
Supplier:  Medchemexpress LLC HY1389610MG
Encompass_Preferred
PD168393 is a potent, selective, cell-permeable irreversible inhibitor of EGFR and ErbB2 tyrosine kinases. It covalently modifies the receptor active site and is used in biochemical and cellular studies to probe EGFR/ErbB2 signaling and inhibition.
  • High potency and selectivity for EGFR and ErbB2.
  • Irreversible covalent inhibitor mechanism for sustained target engagement.
  • Cell-permeable, suitable for cellular assays.
  • High purity (>98% by HPLC) appropriate for research use.
  • Molecular formula C17H13BrN4O; molecular weight 369.22 g·mol⁻¹.
Catalog No. 50-002-62036