Learn More
Medchemexpress LLC N-[4-[(3-chloro-4-fluorophenyl)amino]-7-[[(3S)-tetrahydro-3-furanyl]oxy]-6-quinazolin]-4-(dimethylam | 439081-18-2 | MFCD12407405 | 100.0% | 485.94 | C24H25ClFN5O3 | 250 MG
SDP
Supplier:  Medchemexpress LLC HY10261B250MG
Encompass_Preferred
(E/Z)-Afatinib is a research-grade irreversible epidermal growth factor receptor (EGFR) inhibitor provided as a solid for laboratory use. It is used in cellular and biochemical studies to probe EGFR, HER2, and HER4 signaling pathways and is intended for research applications only.
  • Irreversible inhibitor of EGFR, HER2, and HER4.
  • High purity (reported 99.98%).
  • Molecular weight 485.94 and formula C24H25ClFN5O3.
  • White to off-white solid physical form.
  • Suitable for cellular and biochemical assays.
  • Available in a 250 MG package for lab-scale use.
Catalog No. 50-002-46974