Learn More
Medchemexpress LLC N-[4-[[[(3R,4R)-1-[(2S)-2-amino-1-oxopropyl]-3-methyl-4-piperidinyl]amino]sulfonyl]-1-naphthalenyl] | 850330-18-6 | 98.7% | 508.63 g·mol⁻¹ | C27H32N4O4S | 10MG
SDP
Supplier:  Medchemexpress LLC HY12441610MG
Encompass_Preferred
ML604086 is a selective small-molecule inhibitor of the chemokine receptor CCR8 for in vitro research use. It blocks CCL1 binding to CCR8 and suppresses CCL1-mediated chemotaxis; cellular assays report IC50 ≈ 1.0-1.3 μM. The compound is supplied as a solid powder intended for laboratory research.
Catalog No. 50-002-37210