Learn More
Medchemexpress LLC N-[4-[(4-aminophenyl)thio]phenyl]-4-[[(4-methoxyphenyl)sulfonyl]amino]-butanamide | 791835-21-7 | MFCD07784498 | ≥98.0% | 471.6 g/mol | C23H25N3O4S2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY103661100MG
Encompass_Preferred
BI-6C9 is a small-molecule inhibitor of the BH3-interacting domain (Bid/tBid) used in research to block mitochondrial outer membrane permeabilization and apoptosis signaling. It inhibits tBid (reported Kd ≈ 20 μM) and has been shown to prevent cytochrome c release and AIF nuclear translocation in cell-based studies.
  • Inhibitor of tBid (Kd ≈ 20 μM).
  • Prevents cytochrome c release and mitochondrial outer membrane permeabilization.
  • Used in cell-based apoptosis and neuroprotection studies.
  • High purity (≥98%).
  • Soluble in DMSO and DMF; limited solubility in ethanol and DMSO:PBS (1:1).
  • Crystalline solid; store at -20 °C.
Catalog No. 50-002-45039