Learn More
Medchemexpress LLC N-[4-[(4-aminophenyl)thio]phenyl]-4-[[(4-methoxyphenyl)sulfonyl]amino]-butanamide | 791835-21-7 | MFCD07784498 | 98.5% | 471.59 g·mol⁻¹ | C23H25N3O4S2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10366110MM 1ML
Encompass_Preferred
BI-6C9 is a small-molecule inhibitor of the BH3 interacting domain (Bid) used in research to block tBid-mediated mitochondrial outer membrane permeabilization and downstream apoptotic signaling.
  • 10 mM solution in DMSO, ready for in vitro experiments.
  • Supplied as a 1 mL aliquot for convenient handling.
  • High purity (98.47%) suitable for biochemical assays.
  • Inhibits tBid activity and prevents cytochrome c release from mitochondria.
  • Molecular weight 471.59 g·mol⁻¹; formula C23H25N3O4S2.
Catalog No. 50-002-44588