Learn More
Medchemexpress LLC N-(4-((4-ethylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)-4-methyl-3-((6-methyl-7H-pyrrolo... | 1315330-11-0 | MFCD22572824 | 99.9% | 552.59 g/mol | C29H31F3N6O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY7725110MG
Encompass_Preferred
TAK1/MAP4K2 inhibitor 1 is a small-molecule dual inhibitor of TGFβ-activated kinase 1 (TAK1) and mitogen-activated protein kinase kinase kinase kinase 2 (MAP4K2), supplied as a research-grade reagent. It is characterized by high purity and a defined molecular profile suitable for biochemical and cellular studies.
  • Dual inhibitor of TAK1 and MAP4K2.
  • High purity (approx. 99.9%).
  • Molecular weight 552.59 g/mol and formula C29H31F3N6O2.
  • Supplied as a small-molecule research reagent in a 10 mg quantity.
  • For research use only, not for human use.
Catalog No. 50-002-98486