Learn More
Medchemexpress LLC N-{[4-(5,9-diethoxy-6-oxo-6,8-dihydro-7h-pyrrolo[3,4-g]quinolin-7-yl)-3-methylbenzyl]sulfonyl}-2-(2-methoxyphenyl)aceta | 915191-42-3 | MFCD17166964 | 99.3% | 603.69 g/mol | C32H33N3O7S | 5 MG

Supplier: Medchemexpress LLC HY107945MG
MF498 is a selective, orally active E prostanoid receptor 4 (EP4) antagonist used in research on inflammation and prostaglandin-mediated signaling. It is supplied as a solid for in vitro and in vivo studies and includes solubility and storage guidance.
- Selective and orally active EP4 antagonist.
- High purity (99.3%).
- Molecular weight 603.69 g/mol; formula C32H33N3O7S.
- Soluble in DMSO at ≥ 31 mg/mL for in vitro use.
- Provides example in vivo formulation protocols for dosing.
- Stable as powder when stored at -20°C for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.