Learn More
Medchemexpress LLC N-[4-[6-[4-(trifluoromethyl)phenyl]pyrimidin-4-yl]oxy-1,3-benzothiazol-2-yl]acetamide | 659730-32-2 | MFCD14584859 | 99.2% | 430.4 g·mol⁻¹ | C20H13F3N4O2S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10634100MG
Encompass_Preferred
AMG 517 is a small-molecule antagonist of the TRPV1 (vanilloid) ion channel used to probe TRPV1-mediated signaling and pharmacology. It exhibits sub-nanomolar inhibitory potency and is supplied as a research-grade chemical with batch-specific quality documentation.
  • Potent TRPV1 antagonist with reported IC50 in the sub-nanomolar range.
  • Molecular formula C20H13F3N4O2S and molecular weight 430.4 g·mol⁻¹.
  • High purity (≈99.2% by HPLC) suitable for biochemical assays.
  • Available in solid and solution formats for assay flexibility.
  • Used for in vitro and in vivo pharmacology and target validation.
  • Provided with batch-specific certificate of analysis for QC.
Catalog No. 50-002-42642