Learn More
Medchemexpress LLC N-((4,6-Dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-4'-((4-(7-((2-(2,6-dioxopiperidin-3-yl)-1, 3-d | 2641601-67-2 | MFCD16652626 | 99.4% | 942.15 | C54H67N7O8 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14752510MG
Encompass_Preferred
PROTAC EZH2 Degrader-1 is a research-grade proteolysis-targeting chimera that induces degradation of EZH2 and inhibits its methyltransferase activity. It is intended for in vitro target validation and biochemical studies and is supplied with analytical documentation for research use.
  • Potent EZH2 degradation with IC50 = 2.7 nM.
  • High purity (99.44%) suitable for biochemical assays.
  • Molecular weight 942.15 and formula C54H67N7O8.
  • Supplied in small-mass pack sizes appropriate for laboratory research.
  • Includes accompanying analytical documents: datasheet, COA, HNMR, and LCMS.
Catalog No. 50-002-85912