Learn More
Medchemexpress LLC N-[4-[[8-[(3S)-3,4-dimethyl-1-piperazinyl]-1,5-dihydro-7-methyl-5-oxo-2H-benzopyrano[3,4-c]pyridin-3 | 2227149-22-4 | 99.9% | C28H31F3N4O6S | 10MG
SDP
Supplier:  Medchemexpress LLC HY13025110MG
Encompass_Preferred
DS18561882 is a potent, selective small-molecule inhibitor of methylenetetrahydrofolate dehydrogenase 2 (MTHFD2) used as a biochemical and cellular probe to study mitochondrial one-carbon metabolism. It shows low-nanomolar potency and selectivity, and is supplied as a characterized compound for in vitro and in vivo research applications.
  • Low-nanomolar potency against MTHFD2 (IC50 0.0063 μM).
  • Selective over MTHFD1 (IC50 0.57 μM), supporting target discrimination.
  • Suitable for biochemical and cellular assays as a research probe.
  • Characterized with analytical data and high purity for reproducible results.
  • Available in multiple pack sizes and formats for experimental flexibility.
Catalog No. 50-002-59769