Learn More
Medchemexpress LLC N-(4-aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide | 262368-30-9 | MFCD12457658 | >=98.0% | C14H22N4O | 50 G
Supplier: Medchemexpress LLC HY7569950G
N-(4-aminophenyl)-N-methyl-2-(4-methylpiperazin-1-yl)acetamide is a chemical compound that acts as a reagent for the preparation of deuterated derivatives of nintedanib. It is used in studies related to angiokinase inhibitory, antitumor activity, and pharmacokinetic properties.
- Reagent for preparation of deuterated derivatives of nintedanib
- Useful in angiokinase inhibitory research
- Applied in antitumor activity studies
- Valuable for pharmacokinetic property investigations
- Solid form
- Off-white to pale beige color
- Melting point of 152-154°C