Learn More
Medchemexpress LLC N-[[4-(benzamidocarbamoyl)cyclohexyl]methyl]-2-nitro-4-(trifluoromethyl)benzenesulfonamide | 263849-50-9 | 99.1% | 528.50 | C22H23F3N4O6S | 100 MG

Supplier: Medchemexpress LLC HY107728100MG
S 25585 is a potent, selective neuropeptide Y (NPY) Y5 receptor antagonist used as a research compound in preclinical studies of appetite regulation and NPY signaling. It is supplied as a high-purity solid with documented storage stability for laboratory use.
- High purity (99.1%).
- Molecular weight 528.50.
- Chemical formula C22H23F3N4O6S.
- Solid, white to off-white physical form.
- Documented storage stability at room temperature and in solvent at low temperatures.
- Intended for preclinical NPY receptor and appetite regulation research.
- Available in small-mass packages suitable for laboratory experiments.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.