Learn More
Medchemexpress LLC N-[4-(diethylamino)phenyl]-5-methyl-3-phenylisoxazole-4-carboxamide | 544681-96-1 | 99.2% | 349.43 g/mol | C21H23N3O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10348425MG
Encompass_Preferred
GATA4-NKX2-5-IN-1 is a research-grade small molecule inhibitor that disrupts the transcriptional synergy between GATA4 and NKX2-5, used to study cardiac transcription factor interactions and hypertrophic signaling pathways. The compound is supplied as a solid with high reported purity and is intended for in vitro research applications.
  • Inhibits GATA4-NKX2-5 transcriptional synergy (IC50 = 3 μM).
  • High purity (≈99%) suitable for in vitro research.
  • Supplied in small-molecule quantities for assay and screening use.
  • Molecular formula C21H23N3O2; molecular weight 349.43 g/mol.
Catalog No. 50-002-45359