Learn More
Medchemexpress LLC N-(4-hydroxyphenethyl)pent-4-ynamide | 1694495-59-4 | 99.7% | C13H15NO2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13144210MG
Encompass_Preferred
Alkyne-phenol is an alkyne-containing click-chemistry probe for proximity labeling with ascorbate peroxidase (APEX2). It is more cell wall-permeant than biotin-phenol, improving labeling efficiency in intact cells, and enables identification of APEX-labeled sites for membrane topology assignment and proteomic analysis.
  • Improves APEX-labeling efficiency in intact cells.
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • More cell wall-permeant than biotin-phenol.
  • Aids identification of labeling sites and membrane topology assignment.
  • Available as a solid and as a 10 mM solution in DMSO.
  • High purity (99.7%).
Catalog No. 50-002-61827