Learn More
Medchemexpress LLC N-(4-hydroxyphenyl-2,3,5,6-d4)-acetamide | 64315-36-2 | 99.6% | C8H5D4NO2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY66005S10MM/1ML
Encompass_Preferred
Acetaminophen-d4 is the deuterium-labeled form of Acetaminophen (Paracetamol), a selective cyclooxygenase-2 (COX-2) inhibitor and widely used antipyretic and analgesic agent. It also acts as a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor, serving as a tracer and internal standard for quantitative analysis in various spectroscopic methods.
  • Deuterium-labeled form of acetaminophen
  • Selective cyclooxygenase-2 (COX-2) inhibitor
  • Widely used antipyretic and analgesic agent
  • Potent hepatic N-acetyltransferase 2 (NAT2) inhibitor
  • Used as a tracer in drug development
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes for pharmacokinetic and metabolic studies
Catalog No. 50-004-34205