Learn More
Medchemexpress LLC N-(4-hydroxyphenyl-2,3,5,6-d4)-acetamide | 64315-36-2 | 99.6% | C8H5D4NO2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY66005S100MG
Encompass_Preferred
N-(4-hydroxyphenyl-2,3,5,6-d4)-acetamide is the deuterium labeled form of acetaminophen, also known as paracetamol. It acts as a selective cyclooxygenase-2 (COX-2) inhibitor with an IC50 of 25.8 μM and is a potent hepatic N-acetyltransferase 2 (NAT2) inhibitor. This compound is widely recognized as an antipyretic and analgesic agent, intended for research use only.
  • Deuterium labeled compound
  • Selective cyclooxygenase-2 (COX-2) inhibitor
  • Potent hepatic N-acetyltransferase 2 (NAT2) inhibitor
  • Functions as an antipyretic and analgesic agent
  • Suitable for use as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-34047