Learn More
Medchemexpress LLC N-(4-methylphenyl)maleimide | 1631-28-3 | MFCD00022581 | 98.0% | 187.19 | C11H9NO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1295925MG
Encompass_Preferred
p-Tolylmaleimide (N-(4-methylphenyl)maleimide, CAS 1631-28-3) is a maleimide derivative supplied for research use in biochemical and cell biology studies. It is reported to affect apoptosis and cell-cycle progression in cancer cell models and is available as a purified solid or as DMSO solutions for laboratory use.
  • Provides a maleimide functional group for bioconjugation and reactivity studies.
  • Available as solid or 10 mM DMSO solutions for flexible use.
  • High purity (98.0%) suitable for analytical and biological assays.
  • Molecular formula C11H9NO2; molecular weight 187.19 for accurate stoichiometry.
  • Supplied in small quantities appropriate for screening and synthesis.
Catalog No. 50-002-62084