Learn More
Cayman Chemical N 4ACTAmIdophenylIndomet 10mg
SDP
Supplier:  Cayman Chemical 7027810 MG
Encompass_Preferred
N-4AIA is one of several aromatic amides of indomethacin reported to be potent and selective reversible inhibitors of COX-2.{8243} N-4AIA inhibits human recombinant and ovine COX-2 with IC50 values of 0.1 and 0.625 µM, respectively.{8243,8513} It is about 80 times less potent as an inhibitor of ovine COX-1 than ovine COX-2.
Catalog No. 50-289-5280