Learn More
Medchemexpress LLC N-{5-[3-(3-hydroxypiperidin-1-yl)-1,2,4-oxadiazol-5-yl]-4-methyl-1,3-thiazol-2-yl}acetamide | 1292300-75-4 | 99.0% | 100 MG

Supplier: Medchemexpress LLC HY120438100MG
An orally active PI3Kγ inhibitor for research use with potent PI3Kγ inhibitory activity (IC50 ≈ 29 nM) and additional Akt inhibition (IC50 ≈ 294 nM). The compound is supplied as a white to off-white solid with high purity suitable for laboratory research and is available in multiple pack sizes including 100 MG.
- High purity: 99.0%.
- Molecular formula C13H17N5O3S and molecular weight 323.37 g/mol.
- Physical form: white to off-white solid.
- Potent activity: PI3Kγ IC50 ≈ 29 nM; Akt IC50 ≈ 294 nM.
- Intended for research use only; not for human or veterinary use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.