Learn More
Medchemexpress LLC N-[5-[4-chloro-3-(2-hydroxyethylsulfamoyl)phenyl]-4-methyl-1,3-thiazol-2-yl]acetamide | 593960-11-3 | 99.7% | 1 MG
SDP
Supplier:  Medchemexpress LLC HY120461MG
Encompass_Preferred
PIK-93 is a synthetic small-molecule inhibitor of phosphatidylinositol 4-kinase IIIβ (PI4KIIIβ) with a reported IC50 of 19 nM. It also inhibits class I PI3K isoforms and is used as a research tool to study phosphoinositide signaling, membrane trafficking, and viral replication.
  • Potent PI4KIIIβ inhibitor (IC50 19 nM).
  • Inhibits PI3Kγ and PI3Kα (IC50 16 nM and 39 nM).
  • Soluble in DMSO (100 mg/mL); in vivo formulation protocols provided.
  • Supplied as a white to off-white solid with high purity.
  • Used in cellular assays to probe lipid signaling and viral replication.
Catalog No. 50-002-33261